2-Amino-5-bromo-3-nitropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-nitropyridin-2-amine, 5-Bromo-3-nitro-2-pyridylamine

A316539-100G A316539-25G A316539-5G
77290-228EA 58 USD
77290-228 77290-230 77290-232
2-Amino-5-bromo-3-nitropyridine 95%
2-Amino-5-bromo-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrN₃O₂
MW: 218.01
MDL Number: MFCD00047441
CAS Number: 6945-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR