Dimethyl-2,5-pyridinedicarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl pyridine-2,5-dicarboxylate

A316473-100G A316473-10G A316473-1G A316473-25G A316473-500G A316473-5G
77277-636EA 105 USD
77277-636 77277-638 77277-640 77277-642 77277-644 77277-646
Dimethyl-2,5-pyridinedicarboxylate 98%
Dimethyl 2,5-Pyridinedicarboxylate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD00034767
CAS Number: 881-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR