2-Bromo-4'-(methylsulfonyl)acetophenone 98%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-1-(4-(methylsulphonyl)phenyl)ethanone, 4-(Methylsulphonyl)phenacyl bromide

A316455-1G A316455-25G A316455-5G
77277-630EA 12.36 USD
77277-630 77277-632 77277-634
2-Bromo-4'-(methylsulfonyl)acetophenone 98%
2-Bromo-4'-(methylsulphonyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO₃S
MW: 277.1350 g/mol
MDL Number: MFCD00673134
CAS Number: 50413-24-6
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR