2-Phenylethylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Phenethylboronic Acid Pinacol Ester, 2-Phenylethyl-1-boronic acid pinacol ester

A316032-1G A316032-5G
77277-620EA 22.66 USD
77277-620 77277-622
2-Phenylethylboronic acid pinacol ester 98%
2-Phenylethylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₂
MW: 232.13 g/mol
MDL Number: MFCD03788721
CAS Number: 165904-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR