(S)-5-Benzyl-2-(perfluorophenyl)-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate 95%

Supplier: Ambeed
Danger

A316028-100MG A316028-250MG
77251-062EA 77.25 USD
77251-062 77251-064
(S)-5-Benzyl-2-(perfluorophenyl)-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate 95%
(S)-5-Benzyl-2-(perfluorophenyl)-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₈H₁₃BF₉N₃O
MW: 469.11 g/mol
MDL Number: MFCD32655221
CAS Number: 1327274-33-8
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR