2-Methyl-2H-indazol-5-amine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-methylindazole, 5-Amino-2-methyl-2H-indazole

A316012-100MG A316012-1G A316012-250MG A316012-5G
77263-294EA 20.6 USD
77263-294 77263-296 77263-298 77263-300
2-Methyl-2H-indazol-5-amine 97%
2-Methyl-2H-indazol-5-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.18 g/mol
MDL Number: MFCD03787060
CAS Number: 60518-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR