2-Bromo-1-(5-chloro-2-methoxyphenyl)ethanone 95%

Supplier: Ambeed
Danger

A315698-1G A315698-250MG
77263-290EA 108.15 USD
77263-290 77263-292
2-Bromo-1-(5-chloro-2-methoxyphenyl)ethanone 95%
2-Bromo-1-(5-chloro-2-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MW: 263.516 g/mol
MDL Number: MFCD01793848
CAS Number: 111841-05-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR