DEL-22379 98%

Supplier: Ambeed
Warning

Synonyms: DEL 22379, N-(3-((5-Methoxy-1H-indol-3-yl)methylene)-2-oxoindolin-5-yl)-3-(piperidin-1-yl)propanamide, DEL22379, N-[(3Z)-3-[(5-methoxy-1H-indol-3-yl)methylidene]-2-oxo-1H-indol-5-yl]-3-piperidin-1-ylpropanamide, DEL-22379

A315608-10MG A315608-1MG A315608-25MG A315608-2MG A315608-5MG
77239-484EA 33.99 USD
77239-484 77239-486 77239-488 77239-490 77239-492
DEL-22379 98%
DEL-22379

Precaución: for Research Use Only

Formula: C₂₆H₂₈N₄O₃
MW: 444.53 g/mol
MDL Number: MFCD00950163
CAS Number: 181223-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR