trans-2,6-Dimethylpiperazine dihydrochloride 98%

Supplier: Ambeed
Warning

A315415-100MG A315415-1G A315415-250MG
77263-284EA 66 USD
77263-284 77263-286 77263-288
trans-2,6-Dimethylpiperazine dihydrochloride 98%
trans-2,6-Dimethylpiperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MW: 187.11 g/mol
MDL Number: MFCD16990482
CAS Number: 55116-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR