L-Methionine ethyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Met-OEt·HCl, Ethyl L-methionate hydrochloride

A315362-25G A315362-5G
77277-612EA 27.81 USD
77277-612 77277-614
L-Methionine ethyl ester hydrochloride 98%
L-Methionine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂S·HCl
MW: 213.7 g/mol
MDL Number: MFCD00012508
CAS Number: 2899-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR