4,4'-Didodecyl-2,2'-bithiophene 97%

Supplier: Ambeed
Warning

A315188-1G A315188-5G
77239-480EA 103 USD
77239-480 77239-482
4,4'-Didodecyl-2,2'-bithiophene 97%
4,4'-Didodecyl-2,2'-bithiophene

Precaución: for Research Use Only

Formula: C₃₂H₅₄S₂
MW: 502.90 g/mol
MDL Number: MFCD24539449
CAS Number: 345633-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR