Hoechst 34580 98%

Supplier: Ambeed
Warning

Synonyms: HOE 34580, N,N-Dimethyl-4-(5-(4-methylpiperazin-1-yl)-1H,1'H-[2,5'-bibenzo[d]imidazol]-2'-yl)aniline

A314846-10MG A314846-1MG A314846-25MG A314846-50MG A314846-5MG
77239-468EA 58 USD
77239-468 77239-470 77239-472 77239-474 77239-476
Hoechst 34580 98%
Hoechst 34580

Precaución: for Research Use Only

Formula: C₂₇H₂₉N₇
MW: 451.57 g/mol
MDL Number: MFCD25541584
CAS Number: 23555-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR