4,7-Dibromo-2,3-dihydro-1H-inden-1-one 97%

Supplier: Ambeed
Warning

A314440-100MG A314440-250MG A314440-50MG A314440-1G
77251-024EA 46.35 USD
77251-024 77251-026 77251-028 77746-599
4,7-Dibromo-2,3-dihydro-1H-inden-1-one 97%
4,7-Dibromo-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₆Br₂O
MW: 289.95 g/mol
MDL Number: MFCD22425714
CAS Number: 103515-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR