FH1 (BRD-K4477) 98%

Supplier: Ambeed
Danger

Synonyms: FH1, N,N'-(Methylenebis(4,1-phenylene))diacetamide, N,N′-(Methylenedi-4,1-phenylene)bis-acetamide

A314283-10MG A314283-1MG A314283-5MG
77290-190EA 47 USD
77290-190 77290-192 77290-194
FH1 (BRD-K4477) 98%
FH1 (BRD-K4477)

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₂
MDL Number: MFCD00027653
CAS Number: 2719-05-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR