(-)-DIOP ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R,R)-DIOP, (4R,5R)-(-)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, (((4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene))bis(diphenylphosphine)

A314065-100MG A314065-1G A314065-250MG A314065-25G
77290-182EA 15.45 USD
77290-182 77290-184 77290-186 77290-188
(-)-DIOP ≥98%, ee 99%
(-)-DIOP

Precaución: for Research Use Only

Formula: C₃₁H₃₂O₂P₂
MW: 498.532 g/mol
MDL Number: MFCD00014107
CAS Number: 32305-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR