4-Bromo-2-iodobenzyl bromide 97%

Supplier: Ambeed
Danger

A313844-100MG A313844-250MG A313844-5G
77263-244EA 29 USD
77263-244 77263-246 77263-248
4-Bromo-2-iodobenzyl bromide 97%
4-Bromo-2-iodobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅Br₂I
MDL Number: MFCD18391672
CAS Number: 885681-96-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR