(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate

A313663-100MG A313663-1G A313663-250MG
77251-010EA 78.28 USD
77251-010 77251-012 77251-014
(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥98%, ee 99%
(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₃₆H₁₇F₁₂O₄P
MW: 772.472 g/mol
MDL Number: MFCD08689863
CAS Number: 791616-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR