Pyridine-3,5-diamine 95%

Supplier: Ambeed
Warning

A313469-100MG A313469-1G A313469-250MG A313469-5G
77277-580EA 22.66 USD
77277-580 77277-582 77277-584 77277-586
Pyridine-3,5-diamine 95%
Pyridine-3,5-diamine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.13 g/mol
MDL Number: MFCD00955693
CAS Number: 4318-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR