3-Methylbenzophenone 98%

Supplier: Ambeed
Warning

A312712-1G A312712-25G A312712-5G
77277-568EA 9 USD
77277-568 77277-570 77277-572
3-Methylbenzophenone 98%
3-Methylbenzophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O
MDL Number: MFCD00008535
CAS Number: 643-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR