3-[(Dimethylamino)sulfonyl]benzenecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(N,N-Dimethylsulphamoyl)benzoic acid

A312536-1G A312536-250MG A312536-5G
77277-562EA 59.74 USD
77277-562 77277-564 77277-566
3-[(Dimethylamino)sulfonyl]benzenecarboxylic acid 97%
3-[(Dimethylamino)sulfonyl]benzenecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄S
MW: 229.26 g/mol
MDL Number: MFCD02708231
CAS Number: 7326-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR