4-Amino-N-(4-chlorophenyl)benzenesulfonamide 95%

Supplier: Ambeed
Warning

A312255-100MG A312255-1G A312255-250MG
77290-170EA 49.44 USD
77290-170 77290-172 77290-174
4-Amino-N-(4-chlorophenyl)benzenesulfonamide 95%
4-Amino-N-(4-chlorophenyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₂H₁₁ClN₂O₂S
MDL Number: MFCD02056459
CAS Number: 16803-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR