3,5-Dichloropicolinic acid 95%

Supplier: Ambeed
Warning

A312253-10G A312253-1G A312253-5G
77277-556EA 23 USD
77277-556 77277-558 77277-560
3,5-Dichloropicolinic acid 95%
3,5-Dichloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MDL Number: MFCD03230005
CAS Number: 81719-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR