4'-Methoxy-2-chloroacetophenone 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-1-(4-methoxyphenyl)ethanone, 4-Methoxyphenacyl chloride

A311783-1G A311783-5G
77277-548EA 28.84 USD
77277-548 77277-550
4'-Methoxy-2-chloroacetophenone 98%
4'-Methoxy-2-chloroacetophenone

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.6196 g/mol
MDL Number: MFCD00216508
CAS Number: 2196-99-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR