2,4,6-Tri-tert-butylnitrobenzene 95%

Supplier: Ambeed
Warning

A311601-1G A311601-25G A311601-5G
77250-992EA 22 USD
77250-992 77250-994 77250-996
2,4,6-Tri-tert-butylnitrobenzene 95%
2,4,6-Tri-tert-butylnitrobenzene

Precaución: for Research Use Only

Formula: C₁₈H₂₉NO₂
MDL Number: MFCD00008818
CAS Number: 4074-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR