3-Hydroxy-4-nitrobenzonitrile 98%

Supplier: Ambeed
Danger

A311477-100MG A311477-250MG
77277-544EA 32 USD
77277-544 77277-546
3-Hydroxy-4-nitrobenzonitrile 98%
3-Hydroxy-4-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₂O₃
MDL Number: MFCD06657032
CAS Number: 18495-15-3
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR