3,5-Dimethylcyclohexanone 95% (by GC)

Supplier: Ambeed
Danger

A311341-1G A311341-250MG
77277-534EA 16.48 USD
77277-534 77277-536
3,5-Dimethylcyclohexanone 95% (by GC)
3,5-Dimethylcyclohexanone

Precaución: for Research Use Only

Formula: C₈H₁₄O
MW: 126.1962 g/mol
MDL Number: MFCD00019467
CAS Number: 2320-30-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR