4-Methyl-2-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A311003-100MG A311003-1G A311003-250MG
77277-528EA 31.93 USD
77277-528 77277-530 77277-532
4-Methyl-2-(trifluoromethyl)benzoic acid 98%
4-Methyl-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
MDL Number: MFCD01631591
CAS Number: 120985-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR