4,4'-(Propane-1,2-diyl)bis(piperazine-2,6-dione) 95%

Supplier: Ambeed
Warning

Synonyms: Razoxane

A310864-100MG A310864-250MG A310864-1G
77290-162EA 81.37 USD
77290-162 77290-164 77748-607
4,4'-(Propane-1,2-diyl)bis(piperazine-2,6-dione) 95%
4,4'-(Propane-1,2-diyl)bis(piperazine-2,6-dione)

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₄O₄
MW: 268.27 g/mol
MDL Number: MFCD00430424
CAS Number: 21416-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR