Sivelestat 98%

Supplier: Ambeed
Warning

Synonyms: (2-((4-(Pivaloyloxy)phenyl)sulphonamido)benzoyl)glycine, ONO-5046, 2-(2-(4-(pivaloyloxy)phenylsulphonamido)benzamido)acetic acid

A310710-10MG A310710-1G A310710-250MG A310710-50MG A310710-5MG A310710-1MG
77250-962EA 26.78 USD
77250-962 77250-964 77250-966 77250-968 77250-970 77616-356
Sivelestat 98%
Sivelestat

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O₇S
MW: 434.46 g/mol
MDL Number: MFCD00889071
CAS Number: 127373-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR