N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine 98%

Supplier: Ambeed
Warning

Synonyms: N2,N4,N6-tri-m-tolyl-1,3,5-Triazine-2,4,6-triamine, Tris(3-methylanilino)triazine

A310637-100MG A310637-1G A310637-250MG
77239-420EA 17 USD
77239-420 77239-422 77239-424
N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine 98%
N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine

Precaución: for Research Use Only

Formula: C₂₄H₂₄N₆
MDL Number: MFCD19440763
CAS Number: 82504-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR