2,2'-Dibromo-1,1'-binaphthyl 98%

Supplier: Ambeed
Warning

Synonyms: racemic-2,2'-Dibromo-1,1'-binaphthyl, 2,2-Dibromo-1,1-binaphthyl, 2,2'-Dibromo-1,1'-binaphthalene

A310584-100G A310584-1G
77250-958EA 1299.86 USD
77250-958 77250-960
2,2'-Dibromo-1,1'-binaphthyl 98%
2,2'-Dibromo-1,1'-binaphthyl

Precaución: for Research Use Only

Formula: C₂₀H₁₂Br₂
MW: 412.12 g/mol
MDL Number: MFCD00188005
CAS Number: 74866-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR