3-(tert-Butoxy)-6-chloropyridazine 97%

Supplier: Ambeed
Warning

A310389-10G A310389-1G A310389-250MG A310389-5G
77277-502EA 278.1 USD
77277-502 77277-504 77277-506 77277-508
3-(tert-Butoxy)-6-chloropyridazine 97%
3-(tert-Butoxy)-6-chloropyridazine

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O
MW: 186.6400 g/mol
MDL Number: MFCD03425769
CAS Number: 17321-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR