4-Methoxy-1-naphthaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxynaphthalene-1-carbaldehyde

A310345-1G A310345-25G A310345-5G
77290-156EA 7.21 USD
77290-156 77290-158 77290-160
4-Methoxy-1-naphthaldehyde 97%
4-Methoxy-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
MDL Number: MFCD00004006
CAS Number: 15971-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR