3-Chloro-5-formylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-5-formylphenyl)boronic acid

A310295-100MG A310295-1G A310295-250MG
77277-496EA 39 USD
77277-496 77277-498 77277-500
3-Chloro-5-formylphenylboronic acid 98%
3-Chloro-5-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₃
MDL Number: MFCD09038799
CAS Number: 1451393-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR