(±)-1-Indanol 98%

Supplier: Ambeed
Warning

Synonyms: (±)-1-Hydroxyindan, 1-Indanol, 2,3-Dihydro-1H-inden-1-ol

A310288-100G A310288-10G A310288-25G A310288-5G A310288-500G
77239-412EA 51.5 USD
77239-412 77239-414 77239-416 77239-418 77746-594
(±)-1-Indanol 98%
(±)-1-Indanol

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00003797
CAS Number: 6351-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR