(S)-(+)-1-Boc-3-hydroxypyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

A309934-100G A309934-10G A309934-1G A309934-25G A309934-500G A309934-5G
77263-186EA 55 USD
77263-186 77263-188 77263-190 77263-192 77263-194 77263-196
(S)-(+)-1-Boc-3-hydroxypyrrolidine 97%
(S)-(+)-1-Boc-3-hydroxypyrrolidine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
MW: 187.2362 g/mol
MDL Number: MFCD01317839
CAS Number: 101469-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR