4-(Hydroxymethyl)-3-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A309810-100MG A309810-1G A309810-5G A309810-250MG
77290-144EA 56.65 USD
77290-144 77550-862 77748-604 77748-603
4-(Hydroxymethyl)-3-nitrobenzoic acid 97%
4-(Hydroxymethyl)-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MW: 197.15 g/mol
MDL Number: MFCD01076314
CAS Number: 82379-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR