(S)-3-Hydroxypyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (3S)-pyrrolidin-3-ol hydrochloride

A309806-100G A309806-10G A309806-25G A309806-500G A309806-5G
77277-482EA 104.03 USD
77277-482 77277-484 77277-486 77277-488 77277-490
(S)-3-Hydroxypyrrolidine hydrochloride 97%
(S)-3-Hydroxypyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD00272298
CAS Number: 122536-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR