3-Bromo-6-methoxy-2-methylimidazo[1,2-b]pyridazine 97%

Supplier: Ambeed
Warning

A309569-100MG A309569-1G A309569-250MG
77239-400EA 67.98 USD
77239-400 77239-402 77239-404
3-Bromo-6-methoxy-2-methylimidazo[1,2-b]pyridazine 97%
3-Bromo-6-methoxy-2-methylimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₈H₈BrN₃O
MW: 242.07 g/mol
MDL Number: MFCD22034381
CAS Number: 1369233-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR