1,4-Dithiane-2,5-diol 98% (by NMR)

Supplier: Ambeed
Danger

Synonyms: 2,5-Dihydroxy-1,4-dithiane, p-Dithiane-2,5-diol

A309287-100G A309287-25G
77250-934EA 7.21 USD
77250-934 77250-936
1,4-Dithiane-2,5-diol 98% (by NMR)
1,4-Dithiane-2,5-diol

Precaución: for Research Use Only

Formula: C₄H₈O₂S₂
MW: 152.24 g/mol
MDL Number: MFCD00006659
CAS Number: 40018-26-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR