3-Chloro-9H-carbazole 97%

Supplier: Ambeed
Warning

A309139-1G A309139-250MG A309139-5G
77277-476EA 16 USD
77277-476 77277-478 77277-480
3-Chloro-9H-carbazole 97%
3-Chloro-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₂H₈ClN
MW: 201.65 g/mol
MDL Number: MFCD00222620
CAS Number: 2732-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR