4-Aminotetrahydro-2H-pyran-4-carbonitrile 98%

Supplier: Ambeed
Danger

A309021-250MG A309021-5G A309021-1G
77290-140EA 13.39 USD
77290-140 77290-142 77748-600
4-Aminotetrahydro-2H-pyran-4-carbonitrile 98%
4-Aminotetrahydro-2H-pyran-4-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O
MW: 126.16 g/mol
MDL Number: MFCD08234565
CAS Number: 50289-12-8
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR