Dimethyl fluoromalonate 97%

Supplier: Ambeed
Danger

Synonyms: Dimethyl 2-fluoromalonate

A309020-10G A309020-25G A309020-5G A309020-100G
77290-134EA 6 USD
77290-134 77290-136 77290-138 77616-344
Dimethyl fluoromalonate 97%
Dimethyl fluoromalonate

Precaución: for Research Use Only

Formula: C₅H₇FO₄
MDL Number: MFCD00054680
CAS Number: 344-14-9
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR