Pardoprunox hydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: Pardoprunox HCl, 7-(4-Methylpiperazin-1-yl)benzo[d]oxazol-2(3H)-one hydrochloride

A308996-10MG A308996-1MG A308996-25MG A308996-5MG
77239-386EA 576.8 USD
77239-386 77239-388 77239-390 77239-392
Pardoprunox hydrochloride 98+%
Pardoprunox hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClN₃O₂
MW: 269.72 g/mol
MDL Number: MFCD09833663
CAS Number: 269718-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR