Methyl 3-(chlorosulfonyl)propanoate 95%

Supplier: Ambeed
Danger

A308866-250MG A308866-1G A308866-5G
77277-474EA 9 USD
77277-474 77550-854 77550-856
Methyl 3-(chlorosulfonyl)propanoate 95%
Methyl 3-(chlorosulfonyl)propanoate

Precaución: for Research Use Only

Formula: C₄H₇ClO₄S
MW: 186.614 g/mol
MDL Number: MFCD07339056
CAS Number: 15441-07-3
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR