5-Chloro-2-methoxybenzonitrile 98%

Supplier: Ambeed
Danger

A307990-1G A307990-250MG A307990-25G A307990-5G
77239-376EA 9 USD
77239-376 77239-378 77239-380 77239-382
5-Chloro-2-methoxybenzonitrile 98%
5-Chloro-2-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MDL Number: MFCD00052867
CAS Number: 55877-79-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR