Dipentaerythritol 90%

Supplier: Ambeed
Warning

Synonyms: 2,2'-(Oxybis(methylene))bis(2-(hydroxymethyl)propane-1,3-diol)

A307913-1000G A307913-100G A307913-25G A307913-10G
77250-922EA 35.02 USD
77250-922 77250-924 77250-926 77616-336
Dipentaerythritol 90%
Dipentaerythritol

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₇
MW: 254.28 g/mol
MDL Number: MFCD00004691
CAS Number: 126-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR