Amcasertib 97%

Supplier: Ambeed
Warning

Synonyms: N-(2-(Diethylamino)ethyl)-2,4-dimethyl-5-((2-oxo-5-(2-phenylthiazol-4-yl)indolin-3-ylidene)methyl)-1H-pyrrole-3-carboxamide

A307810-10MG A307810-1MG A307810-25MG A307810-50MG A307810-5MG
77250-912EA 50.47 USD
77250-912 77250-914 77250-916 77250-918 77250-920
Amcasertib 97%
Amcasertib

Precaución: for Research Use Only

Formula: C₃₁H₃₃N₅O₂S
MW: 539.69 g/mol
MDL Number: MFCD30489231
CAS Number: 1129403-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR