2-Bromobenzo[b]naphtho[2,3-d]furan 97%

Supplier: Ambeed
Warning

A307675-10G A307675-1G
77239-360EA 49 USD
77239-360 77239-362
2-Bromobenzo[b]naphtho[2,3-d]furan 97%
2-Bromobenzo[b]naphtho[2,3-d]furan

Precaución: for Research Use Only

Formula: C₁₆H₉BrO
MDL Number: MFCD30187285
CAS Number: 1627917-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR