1-(1H-1,3-Benzodiazol-2-yl)ethan-1-one 96%

Supplier: Ambeed
Warning

Synonyms: 1-(1H-Benzo[d]imidazol-2-yl)ethanone, 1-(1H-1,3-Benzimidazol-2-yl)-1-ethanone

A307673-100MG A307673-1G A307673-250MG A307673-25G A307673-5G
77263-158EA 7 USD
77263-158 77263-160 77263-162 77263-164 77263-166
1-(1H-1,3-Benzodiazol-2-yl)ethan-1-one 96%
1-(1H-1,3-Benzodiazol-2-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD00159994
CAS Number: 939-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR